Benzenesulphonanilide ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:1678-25-7
:
Formula:C₁₂H₁₁NO₂S
MDL Number:MFCD00035919
:233.29 g/mol
EINECS:216-832-4
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:108.0 to 112.0 deg-C
- Solubility in Methanol:almost transparency