Order Entry
Sweden
ContactUsLinkComponent
Dibutylammonium acetate ∼0.5 mol/L in water for ion pair chromatography, for LC-MS
Dibutylammonium acetate ∼0.5 mol/L in water for ion pair chromatography, for LC-MS
Supplier:  TCI
Certificates
undefined
Dibutylammonium acetate ∼0.5 mol/L in water for ion pair chromatography, for LC-MS
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless clear liquid
  • Concentration:
    0.50 to 0.53 mol/L
  • Absorbance (230nm):
    max. 1.600
  • Absorbance (254nm):
    max. 0.050
  • Absorbance (270nm):
    max. 0.020

Specification Test Results

Related Information