(S)-(-)-DBD-Pro-COCl [=(S)-(-)-4-(N,N-Dimethylaminosulphonyl)-7-(2-chloroformylpyrrolidin-1-yl)-2,1,3-benzoxadiazole] ≥90.0% (by HPLC) HPLC labeling reagent for e.e. determination
Catalog # TCIAA5564-100MG
Leverantör: TCI
CAS-nummer:
Specifikationer
- Pk:100 mg
- Seq:2440
Specifikationer
Specifikationstestresultat
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (HPLC):min. 90.0 area%
- Optical purity (LC):min. 99.0 ee%
- Elemental analysis (Nitrogen):15.00 to 16.00 %
- Effect test of HPLC derivatization:Effective as labeling agent fo 2-Hexanol
Specifikationstestresultat
Om denna produkt
:
:358.80 g/mol
:GHS07
Förvaringstemperatur:Refrigerator
CAS-nummer:150993-63-8
Formel:C₁₃H₁₅ClN₄O₄S
MDL-nummer:MFCD00191380