Order Entry
Sweden
ContactUsLinkComponent
 
5-Amino-1-(2,6-dichloro-4-(trifluoromethyl)phenyl)-1H-pyrazole-3-carbonitrile ≥98.0% (by GC)
 :  TCI
 
5-Amino-1-(2,6-dichloro-4-(trifluoromethyl)phenyl)-1H-pyrazole-3-carbonitrile ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Yellow powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    149.0 to 153.0 deg-C