4-Acetylbenzenesulphonamide ≥98.0% (by HPLC, total nitrogen)
TCIAA3136-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:178.0 to 182.0 deg-C
:
:199.23 g/mol
:216-363-5
:GHS07
:1565-17-9
:C₈H₉NO₃S
:MFCD00792524