Azido-PEG2-Amine.Tos-OH ≥95.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:2173092-98-1
:
Formula:C₆H₁₄N₄O₂·C₇H₈O₃S
:346.40 g/mol
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 95.0 area%
- Elemental analysis (Nitrogen):15.30 to 17.00 %