(-)-Arctiin ≥97.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:20362-31-6
:
Formula:C₂₇H₃₄O₁₁
MDL Number:MFCD00870630
:534.6 g/mol
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 97.0 area%
- Melting point:107.0 to 111.0 deg-C
- Specific rotation [a]20/D:-50.0 to -55.0 deg (C=2, MEOH)