
2-Aminobenzo[d]thiazol-4-ol ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:7471-03-6
:
Formula:C₇H₆N₂OS
MDL Number:MFCD00160060
:166.2003 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:182.0 to 186.0 deg-C