Order Entry
Sweden
ContactUsLinkComponent
 
 
4-[(6-Acryloyloxy)hexyloxy]-4'-cyanobiphenyl ≥98.0% (by GC)
 :  TCI
 
undefined
 
4-[(6-Acryloyloxy)hexyloxy]-4'-cyanobiphenyl ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %