Acrylamide ∼50% in water
Supplier: TCI
Certificates
About this item
CAS Number:79-06-1
:
Formula:C₃H₅NO
MDL Number:MFCD00008032
:71.08 g/mol
Merck Index:14,00129
Storage Temperature:Refrigerator
EINECS:201-173-7
UN:3426
ADR:6.1,III
:GHS06
:GHS08
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Concentration (Bromination Method):49.0 to 51.0 w/w%