4-Amylcyclohexylamine (cis- and trans- mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
Catalog # TCIAA2525-5ML
Supplier: TCI
CAS Number:
Specifications
- Pk:5 mL
- Seq:2210
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 % (total of isomers)
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
About this item
:
:169.31 g/mol
Boiling Pt:76°C
UN:2735
ADR:8,III
:GHS05
CAS Number:38793-01-0
Formula:C₁₁H₂₃N
MDL Number:MFCD07778407