(S)-(-)-2-Amino-3,3-dimethyl-1,1-diphenyl-1-butanol ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:144054-70-6
:
Formula:C₁₈H₂₃NO
MDL Number:MFCD03427198
:269.39 g/mol
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 95.0 %
- Melting point:132.0 to 136.0 deg-C
- Specific rotation [a]20/D:-144 to -150 deg (C=0.4, EtOH)