
Adiphenine hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:50-42-0
:
Formula:C₂₀H₂₅NO₂
MDL Number:MFCD00050508
:347.88 g/mol
Merck Index:14,00161
EINECS:200-036-9
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:115.0 to 119.0 deg-C