Adefovir dipivoxil ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:142340-99-6
:
Formula:C₂₀H₃₂N₅O₈P
MDL Number:MFCD00869897
:501.48 g/mol
Merck Index:14,00151
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:99.0 to 103.0 deg-C