
5-Amino-2-methylbenzoic acid ≥98.0% (by titrimetric analysis)
: TCI
:2840-04-2
:
:C₈H₉NO₂
:MFCD06208351
:151.16 g/mol
:GHS07
:Refrigerator
- Appearance:White to Light yellow to Light orange powder to crystalline
- Purity (HPLC):min. 97.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:195.0 to 199.0 deg-C