5-Acetyl-3-chloro-10,11-dihydrodibenzo[b,f]azepine ≥98.0% (by GC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:25961-11-9
:
Formula:C₁₆H₁₄ClNO
MDL Number:MFCD01632170
:271.74 g/mol
EINECS:247-371-7
Specification Test Results
- Appearance:White to Orange to Green powder to crystalline
- Purity (GC):min. 98.0 %
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:122.0 to 126.0 deg-C