D-Alanine methyl ester hydrochloride ≥97.0% (by total nitrogen and titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:14316-06-4
:
Formula:C₄H₉NO₂
MDL Number:MFCD00066141
:139.58 g/mol
EINECS:238-258-3
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (with Total Nitrogen):min. 97.0 %
- Purity (Argentmetric Titration):min. 97.0 %
- Specific rotation [a]20/D:-6.0 to -9.0 deg (C=2, MeOH)