Acesulphame potassium ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:55589-62-3
:
Formula:C₄H₄KNO₄S
MDL Number:MFCD00043833
:201.24 g/mol
Merck Index:14,00037
EINECS:259-715-3
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Ion exchange titration):min. 98.0 %
- Solubility in Water:almost transparency