(R)-2-Acetoxy-2-phenylacetic acid ≥98.0% (by GC, titration analysis)
: TCI
:51019-43-3
:
:C₁₀H₁₀O₄
:MFCD00004249
:194.19 g/mol
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:98.0 to 102.0 deg-C
- Specific rotation [a]20/D:-151 to -155 deg (C=2, Aceton)