Order Entry
Sweden
ContactUsLinkComponent
n-Octyl acetoacetate ≥95.0% (by GC)
Irritant
n-Octyl acetoacetate ≥95.0% (by GC)
Catalog # TCIAA0915-25ML
Supplier:  TCI
CAS Number:  
Irritant
n-Octyl acetoacetate ≥95.0% (by GC)
Catalog # TCIAA0915-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results

About this item

 :
 :214.31 g/mol
Boiling Pt:198°C
EINECS:240-491-0
 :GHS07
CAS Number:16436-00-3
Formula:C₁₂H₂₂O₃
MDL Number:MFCD00059448