Aniline-2,5-disulphonic acid monosodium salt ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:24605-36-5
:
Formula:C₆H₇NO₆S₂
MDL Number:MFCD00078393
:275.22 g/mol
EINECS:246-349-4
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Solubility in hot Water:almost transparency