Order Entry
Sweden
ContactUsLinkComponent
beta-Alanyl-L-histidine
beta-Alanyl-L-histidine
Supplier:  TCI
Certificates
beta-Alanyl-L-histidine
Supplier:  TCI

Specifications

  • CAS No.:
    305-84-0
  • Form:
    White powder to crystal
  • MDL No.:
    MFCD00005207
  • Protein synonyms:
    L-Ignotine|L-Carnosine
  • Protein/peptide name:
    β-Alanyl-L-histidine
  • Purity:
    >98,0% (HPLC,T)
  • Molecular weight:
    226,24 g/mol
  • Molecular formula:
    C₉H₁₄N₄O₃

Specifications

Related Information