Order Entry
Sweden
ContactUsLinkComponent
Sodium hydroxide 3 mol/l (3 N) in aqueous solution
 
Sodium hydroxide 3 mol/l (3 N) in aqueous solution
  5657.5000
 :  
undefined
 
Sodium hydroxide 3 mol/l (3 N) in aqueous solution
  5657.5000
 :  

 

  • Pack type:
    Plastic container
  • Pk:
    5 L
  • Seq:
    5190

 

 

 :
 :40 g/mol
 :109 °C (1013 hPa)
 :≤1.11 g/cm³ (20 °C)
 :Ambient
 :215-185-5
 :1824
 :8,II
 :GHS05
 :1310-73-2
 :NaOH
 :MFCD00003548