Sodium hypochlorite (3.5% Cl₂) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7681-52-9
:
Formula:ClNaO
MDL Number:MFCD00011120
:74.44 g/mol
Storage Temperature:Refrigerator
EINECS:231-668-3
Specification Test Results
- Assay (Active chlorine)(at filling):3.5 - 5.0 %
- Alkalinity:Max. 0.05 meq/g
- Chlororometric degrees (Fr;at packaging):11.1 - 15.8 °
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 5L Plastic drum DIN 51 UN (ZPACK13951001)
Förpackningsinformation
27896_SODIUM HYPOCHLORITE 3,5 % ACT CL GPR REC_BSE_REV0.pdf
BSE/TSE-certifikat
SDS_EN_CH_VWRC27896.pdf SDS_EN_DE_VWRC27896.pdf SDS_EN_NL_VWRC27896.pdf SDS_EN_UK_VWRC27896.pdf sds_VWRC27896
SDS
spec_VWRC27896.291 spec_VWRC27896.360