Sodium hypochlorite (14% Cl₂) in aqueous solution, TECHNICAL
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7681-52-9
:
Formula:ClNaO
MDL Number:MFCD00011120
:74.44 g/mol
Boiling Pt:111 °C (1013 hPa)
Melting Pt:–30…–20 °C
Density:1.22 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-668-3
UN:1791
ADR:8,II
:GHS05
:GHS09
Specification Test Results
- Assay (Active chlorine)(W/V)(at filling):Min. 11.5 %
- Identification:Passes test