Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium chloride
Sodium chloride 0.9% in aqueous solution, sterile
Sodium chloride 0.9% in aqueous solution, sterile
:
PanReac AppliChem
Sodium chloride 0.9% in aqueous solution, sterile
:
PanReac AppliChem
:
7647-14-5
:
:
NaCl
:
MFCD00003477
:
58.44 g/mol
:
Ambient
:
231-598-3
pH (20 °C):
5,0 - 7,5
APLIA1671.0100_TDS_EN
APLIA1671.0250_TDS_EN