Fehling's reagent II (L(+)-potassium sodium tartrate in sodium hydroxide solution)
Leverantör: Thermo Fisher Scientific
Certifikat
Om denna produkt
CAS-nummer:6381-59-5
MDL-nummer:MFCD00065391
Kokande Pt:100 °C (760 mmHg)
Densitet:1.24 g/cm³
Förvaringstemperatur:Ambient
FN:1824
ADR:8,II
:GHS05
Specifikationstestresultat
- Appearance:Clear colorless Solution
- Solution contains:346 g C4H4O6KNa.4H2O + 100g NaOH/l H2O