Order Entry
Sweden
ContactUsLinkComponent
Formaldehyde 4% stabilised, buffered (pH 7.0 ±0.1)
Health
Irritant
Formaldehyde 4% stabilised, buffered (pH 7.0 ±0.1)
Supplier:  VWR Chemicals
Certificates
undefined
Health
Irritant
Formaldehyde 4% stabilised, buffered (pH 7.0 ±0.1)
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (Formaldehyde):
    3.5 - 4.5 %
  • pH (20 °C):
    6.9 - 7.1
  • Methanol:
    0.5 - 1.5 %

Specification Test Results

Frequently Bought Together