Order Entry
Sweden
ContactUsLinkComponent
Orthophosphoric acid 150 ml/l 150 ml/l, AVS TITRINORM for TOC
Corrosive
Orthophosphoric acid 150 ml/l 150 ml/l, AVS TITRINORM for TOC
Catalog # 310337.2500
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Orthophosphoric acid 150 ml/l 150 ml/l, AVS TITRINORM for TOC
Catalog # 310337.2500
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

About this item

 :
 :98 g/mol
Density:1.1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-633-2
UN:1805
ADR:8,III
 :GHS05
CAS Number:7664-38-2
Formula:H₃PO₄
MDL Number:MFCD00011340