Order Entry
Sweden
Orders LinkContactUsLinkComponent
Sodium alginate, TECHNICAL

Sodium alginate, TECHNICAL

Plastic bottle for solids, 1 kg
Catalog # 27660.296
Supplier:   VWR Chemicals
Sodium alginate, TECHNICAL
Catalog # 27660.296
Supplier:  VWR Chemicals
CAS Number:  9005-38-3
Sodium alginate, TECHNICAL
Sodium alginate, TECHNICAL

Plastic bottle for solids, 1 kg

CAS Number:  9005-38-3
Price
4 825,00 kr
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Alginic acid sodium salt, Algin
  • CAS Number:9005-38-3
  • MDL Number:MFCD00081310
  • Storage Temperature:Ambient
Specifications

Specification Test Results

  • Identification:
    Passes test

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg

Specifications