Order Entry
Sweden
ContactUsLinkComponent
Copper (II) ethylenediamine complex 1 mol/l (3 N) in aqueous solution, TECHNICAL
Corrosive
Irritant
Copper (II) ethylenediamine complex 1 mol/l (3 N) in aqueous solution, TECHNICAL
Catalog # 24991.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Irritant
Copper (II) ethylenediamine complex 1 mol/l (3 N) in aqueous solution, TECHNICAL
Catalog # 24991.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Density (20/4):
    1.096 - 1.100
  • Mol proportion (Ethylenediamine/Cu):
    1.96 - 2.04

Specification Test Results

About this item

 :
 :217.76 g/mol
Density:1.1 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1761
ADR:8 (6.1),II
 :GHS05
 :GHS07
CAS Number:14552-35-3
 :13426-91-0
Formula:Cu(H₂NCH₂CH₂NH₂)₂(OH)₂
MDL Number:MFCD00274628