Order Entry
Sweden
ContactUsLinkComponent
Anti-IgG Goat Antibody (CyDye™ 700)
Anti-IgG Goat Antibody (CyDye™ 700)
Supplier:  Cytiva
Certificates
undefined
Anti-IgG Goat Antibody (CyDye™ 700)
Supplier:  Cytiva

Specifications

  • Catalog No:
  • 29-3607-85
  • 29-3607-84
  • Antibody type:
  • Secondary
  • Secondary
  • Conjugation:
  • CyDye™ 700
  • CyDye™ 700
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Goat
  • Goat
  • Western blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • 2...8 °C
  • 2...8 °C

Specifications

Related Information