Order Entry
Sweden
ContactUsLinkComponent
Human Recombinant FGF-4 (Animal free) (from E. coli)
Human Recombinant FGF-4 (Animal free) (from E. coli)
Certificates
undefined
Human Recombinant FGF-4 (Animal free) (from E. coli)

Specifications

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0,1
  • Biological activity:
    Bioactivity assay: NR6R-3T3 Proliferation, ED50 ≤5 ng/ml, Bioactivity ≥2,0×10⁵ units/mg
  • Form:
    Lyophilised
  • Gene ID:
    P08620
  • Reconstitution instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    Y
  • Protein synonyms:
    Transforming protein KS3|HST-1|HBGF-4
  • Protein/peptide name:
    FGF-4
  • Purity:
    ≥95%
  • Molecular weight:
    19,4 kDa
  • Sequence:
    MAPTAPNGTL EAELERRWES LVALSLARLP VAAQPKEAAV QSGAGDYLLG IKRLRRLYCN VGIGFHLQAL PDGRIGGAHA DTRDSLLELS PVERGVVSIF GVASRFFVAM SSKGKLYGSP FFTDECTFKE ILLPNNYNAY ESYKYPGMFI ALSKNGKTKK GNRVSPTMKV THFLPRL
  • Endotoxin level:
    Low
  • Formulation:
    20 mM sodium phosphate, 0,5 M sodium chloride, pH 7,5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

Specifications