Acrylamide : Bisacrylamide (37.5:1) 30% in water for electrophoresis
TCIAA3218-250ML
: TCI
:
- Pk:250 mL
- Seq:2590
- Appearance:Colorless clear liquid
- Functionality test:Electrophoresis to pass test
:
:Refrigerator
:3426
:6.1,III
:GHS08
:GHS07
:79-06-1
:MFCD00080848