Order Entry
Sweden
ContactUsLinkComponent
Acrylamide : Bisacrylamide (29:1) 30% in water for electrophoresis
Acrylamide : Bisacrylamide (29:1) 30% in water for electrophoresis
Catalog # TCIAA3217-250ML
Supplier:  TCI
CAS Number:  
Acrylamide : Bisacrylamide (29:1) 30% in water for electrophoresis
Catalog # TCIAA3217-250ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    250 mL
  • Seq:
    2590

Specifications

Specification Test Results

  • Appearance:
    Colorless clear liquid
  • Functionality test:
    Electrophoresis to pass test

Specification Test Results

About this item

 :
Storage Temperature:Refrigerator
UN:3426
ADR:6.1,III
 :GHS08
 :GHS07
 :79-06-1
MDL Number:MFCD00080848