Industrial gloves, phynomic airLite A ESD
Supplier: UVEX
Certificates
About this item
The most lightweight safety glove within its range with ESD-Function according to DIN 16350.
- An excellent combination of high dexterity, touch sensitivity, lightweight design, and breathability
- Aqua-polymer foam airLite coating on palm and fingertips
- Outer material made from polyamide, elastane, and carbon
- Use on almost all screens, tablets, and mobile phones
- Excellent grip in dry and slightly damp areas
The new Aqua-Polymer foam airLite coating, together with a very thin liner (18 gauge), provides very good dexterity for precision work and handling of very small pieces, in dry and slightly damp areas. The additional touchscreen ability make the phynomic airLite A suitable on all touchscreens, tablets and mobile phones. These safety glove are free of harmful substances, with a skin compatibility that is dermatological approved (proDerm). The gloves are therefore also very useful for allergic persons.
Certifications: EN 388 (3110X), DIN EN 16350:2014
Packaging: 10 pairs per bundle.
Specifications
- Catalog No:
- UVEX6003808
- UVEX6003809
- UVEX6003810
- UVEX6003811
- UVEX6003812
- UVEX6003806
- UVEX6003807
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Liner material:
- Polyamide/elastane/carbon
- Polyamide/elastane/carbon
- Polyamide/elastane/carbon
- Polyamide/elastane/carbon
- Polyamide/elastane/carbon
- Polyamide/elastane/carbon
- Polyamide/elastane/carbon
- Finish:
- Coated fingertips/palm
- Coated fingertips/palm
- Coated fingertips/palm
- Coated fingertips/palm
- Coated fingertips/palm
- Coated fingertips/palm
- Coated fingertips/palm
- Gauge:
- 18
- 18
- 18
- 18
- 18
- 18
- 18
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388 (3110X), DIN EN 16350:2014
- EN 388 (3110X), DIN EN 16350:2014
- EN 388 (3110X), DIN EN 16350:2014
- EN 388 (3110X), DIN EN 16350:2014
- EN 388 (3110X), DIN EN 16350:2014
- EN 388 (3110X), DIN EN 16350:2014
- EN 388 (3110X), DIN EN 16350:2014
- Anti-static:
- +
- +
- +
- +
- +
- +
- +