Methyl Acetate, standard material for GC
Catalog # TCIAS0300-5ML
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 mL
- CAS No.:79-20-9
- Description:Methyl Acetate, Purity (GC): min. 99.5 %
- Formula:C₃H₆O₂
- Description:Methyl Acetate
- Flash point:–16 °C (lit.)
- Boiling point:57 °C (lit.)
- Appearance:Colorless clear liquid