Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Food Standards
Food Red No. 40
Food Red No. 40
Food Red No. 40
TCIAF0325-25G
:
TCI
:
Food Red No. 40
TCIAF0325-25G
:
TCI
:
Pk:
25 g
CAS No.:
25956-17-6
Description:
Food Red No. 40
Formula:
C₁₈H₁₄N₂Na₂O₈S₂
Description:
Food Red No. 40
Colour Index 16035