Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Food Standards
Food Red No. 104
Food Red No. 104
Food Red No. 104
Catalog # TCIAF0142-25G
Supplier:
TCI
CAS Number:
Expand image
Food Red No. 104
Catalog # TCIAF0142-25G
Supplier:
TCI
CAS Number:
Specifications
Pk:
25 g
CAS No.:
18472-87-2
Description:
Food Red No. 104
Formula:
C₂₀H₂Br₄Cl₄Na₂O₅
Description:
Food Red No. 104
Specifications
About this item
Product Details
Documentation
Product Details
Documentation