Order Entry
Sweden
ContactUsLinkComponent
Food Red No. 102
Food Red No. 102
Catalog # TCIAF0140-25G
Supplier:  TCI
CAS Number:  
undefined
Food Red No. 102
Catalog # TCIAF0140-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • CAS No.:
    2611-82-7
  • Description:
    Food Red No. 102
  • Formula:
    C₂₀H₁₁N₂Na₃O₁₀S₃
  • Description:
    Food Red No. 102

Specifications

About this item