Food Green No. 3
Catalog # TCIAF0146-10G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- CAS No.:2353-45-9
- Description:Food Green No. 3
- Formula:C₃₇H₃₄N₂Na₂O₁₀S₃
- Description:Food Green No. 3
Specifications
About this item
Colour index 42053
Form: Powder
Color: Green