0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
Standards
Food Standards
Food Blue No. 1
Food Blue No. 1
Catalog # TCIAF0147-25G
Supplier:
TCI
CAS Number:
Expand image
Food Blue No. 1
Catalog # TCIAF0147-25G
Supplier:
TCI
CAS Number:
Specifications
Pk:
25 g
CAS No.:
3844-45-9
Description:
Food Blue No. 1
Formula:
C₃₇H₃₄N₂Na₂O₉S₃
Description:
Food Blue No. 1
Specifications
About this item
Product Details
Documentation
Product Details
Documentation