Aluminium foil tapes, 1436
: 3M
3M™ aluminum foil tape 1436 is a high-quality tape for internal and external ducting work. It offers an aggressive adhesive for ease of application.
- Foil backing provides an excellent reflective surface for both heat and light
- Low water vapour transmission rate offers excellent sealing and patching performance
- Performance range: –25 to 70 °C (short term 100 °C)
- Very high initial tack to most surfaces
- Easy release liner
- Tensile strength: 45 [Newton/25 mm]
- Dead soft aluminium back material
It has an aggressive rubber adhesive and is supplied with easy release liner and a soft aluminium backing, which provides good conformability.
This tape can be used in internal and external applications. It bonds to a variety of different surfaces including woods, metals, and plastics.
: Meets low flammable rating according to DIN 4102B1.