Order Entry
Sweden
ContactUsLinkComponent
 
 
 
 
Toluene, anhydrous ≥99.9% for applications requiring low water content <0.0010% H₂O, Riedel-de Haën™
  HONC66212-100ML
 :  
undefined
 
 
 
Toluene, anhydrous ≥99.9% for applications requiring low water content <0.0010% H₂O, Riedel-de Haën™
  HONC66212-100ML
 :  

 

  • Pack type:
    Bottle, septum-sealed
  • Pk:
    100 ml
  • Seq:
    2440

 

 

  • Assay (GC):
    Min. 99.9 %
  • Water (Karl Fischer):
    Max. 0.0010 %
  • Non-volatile matter:
    Max. 0.0001 %

 

 

 :
 :92.14 g/mol
 :110.6 °C
 :–95 °C
 :0.867 g/cm3 (20 °C)
 :4 °C
 :Ambient
 :203-625-9
 :1294
 :3,II
 :Packed in septum-sealed bottle.
 :GHS07
 :GHS08
 :GHS02
 :108-88-3
 :C₆H₅CH₃
 :MFCD00008512