Order Entry
Sweden
ContactUsLinkComponent
Boron trifluoride - methanol complex solution 13-15% (BF₃ basis)
Corrosive
Health
Toxic
Flammable
Boron trifluoride - methanol complex solution 13-15% (BF₃ basis)
Catalog # HONC61626H-500ML
CAS Number:  
Corrosive
Health
Toxic
Flammable
Boron trifluoride - methanol complex solution 13-15% (BF₃ basis)
Catalog # HONC61626H-500ML
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Assay of BF₃:
    13 - 15 %

Specification Test Results

About this item

 :
 :99.85 g/mol
Boiling Pt:64 °C
Density:0.880 g/cm3
Flash Pt:11 °C
UN:3286
ADR:3,II
 :GHS05
 :GHS08
 :GHS06
 :GHS02
CAS Number:373-57-9
 :16045-88-8
Formula:CH₄BF₃O
MDL Number:MFCD00071635