(S)-Trolox methyl ether ≥98.0%, LiChropur™ for chiral derivatisation, Supelco®
Catalog # 93510-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Seq:2370
Specifications
About this item
:
:264.32 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:135806-59-6
Formula:C₁₅H₂₀O₄
MDL Number:MFCD00171453