Order Entry
Sweden
ContactUsLinkComponent
 
Nα-(2,4-Dinitro-5-fluorophenyl)-D-valinamide ≥98.0%, LiChropur™ for chiral derivatisation, Supelco®
Catalog # 42100-500MG
Supplier:  Merck
CAS Number:  
undefined
Nα-(2,4-Dinitro-5-fluorophenyl)-D-valinamide ≥98.0%, LiChropur™ for chiral derivatisation, Supelco®
Catalog # 42100-500MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :300.24 g/mol
 :M-Clarity™ quality level = MQ100
 :GHS07
Storage Temperature:Refrigerator
CAS Number:210529-62-7
Formula:C₁₁H₁₃FN₄O₅
MDL Number:MFCD00155643