Order Entry
Sweden
ContactUsLinkComponent
Ethyl-beta-D-glucuronide-(ethyl-d5), certified reference material, Cerilliant®, Supelco®
Ethyl-beta-D-glucuronide-(ethyl-d5), certified reference material, Cerilliant®, Supelco®
  E-063-1ML
 :  Merck
 :  
undefined
Ethyl-beta-D-glucuronide-(ethyl-d5), certified reference material, Cerilliant®, Supelco®
  E-063-1ML
 :  Merck
 :  

 

  • Concentration:
    1,0 mg/ml in methanol
  • Pk:
    1 mL
  • Standards:
    Certified reference material
  • CAS No.:
    1135070-98-2
  • Description:
    Ethyl-beta-D-glucuronide-(ethyl-d5)
  • Formula:
    C₈D₅H₉O₇
  • Description:
    Ethyl-beta-D-glucuronide-(ethyl-d5)
  • Pk Info:
    Ampoule of 1 ml

 

 

Ethyl ß-D-glucuronide is a primary urinary metabolite of alcohol and can be used as a biomarker for the monitoring of alchohol consumption.

This internal standard is suitable for quantitation of alcohol levels in the body by LC/MS or GC/MS for applications such as forensic testing, clinical toxicology, or alcohol monitoring.

 : Snap-N-Spike®/Snap-N-Shoot®.