0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
Amberlite® XAD4
Amberlite® XAD4, Pore size: 100 Å 20 - 60 mesh ion-exchange resin, Supelco®
Amberlite® XAD4, Pore size: 100 Å 20 - 60 mesh ion-exchange resin, Supelco®
Amberlite® XAD4, Pore size: 100 Å 20 - 60 mesh ion-exchange resin, Supelco®
Catalog # 10358.
Supplier:
Merck
CAS Number:
Expand image
Amberlite® XAD4, Pore size: 100 Å 20 - 60 mesh ion-exchange resin, Supelco®
Catalog # 10358.
Supplier:
Merck
CAS Number:
Specifications
Pk:
1
Seq:
0001
Specifications
About this item
Product Details
:
CAS Number:
9003-69-4
:
37380-42-0
MDL Number:
MFCD00132704
Product Details