Alizarin red S (sodium salt), AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent (C.I. 58005)
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:130-22-3
:93982-72-0
Formula:C₁₄H₇NaO₇S·1H₂O
MDL Number:MFCD00013049
:342.26 g/mol (anhy)
Specification Test Results
- pH Range (Yellow to Purple-Red):3.7 - 5.2
- Identity (IR-Spectrum):Passes Test
- Insoluble in Water:Passes Test
- Absorption Maximum λ1 (NaOH 0.1 M):592 - 596
- Absorption Maximum λ2 (NaOH 0.1 M):553 - 558
- Absorptivity (A1%/1 cm; λ max. 1, NaOH 0.1 M):305 - 480
- Absorptivity (A1%/1 cm; λ max. 2, NaOH 0.1 M):330 - 520
- Related Substances (TLC):Passes Test
- Loss on Drying (135 °C):≤5.0
- Suitability for Microscopy:Passes Test
- Sensitivity to Aluminium:Passes Test